missing translation for 'onlineSavingsMsg'
Få mere at vide

trans 4-Dimethylaminocrotonic Acid-d6 Methyl Ester, TRC

Artikelnummer. 30375214
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30375214 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375214 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4466372.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Kemisk navn eller materiale trans 4-Dimethylaminocrotonic Acid-d6 Methyl Ester
CAS 1259680-82-4
Molekylær formel C7H7D6NO2
Mængde 2.5 mg
Synonym (E)-4-(Dimethylamino)but-2-enoic Acid-d6 Methyl Ester, (E)-4-(Dimethylamino)-2-butenoic Acid-d6 Methyl Ester
InChI formel InChI=1S/C7H13NO2/c1-8(2)6-4-5-7(9)10-3/h4-5H,6H2,1-3H3/b5-4+/i1D3,2D3
SMIL [2H]C([2H])([2H])N(C\C=C\C(=O)OC)C([2H])([2H])[2H]
IUPAC navn methyl (E)-4-[bis(trideuteriomethyl)amino]but-2-enoate
Molekylvægt (g/mol) 149.22
Formel vægt 149.1323
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.