Få mere at vide
trans 4-Dimethylaminocrotonic Acid-d6 Methyl Ester, TRC
Artikelnummer. 30376985
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30376985 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376985 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44663725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans 4-Dimethylaminocrotonic Acid-d6 Methyl Ester
CAS 1259680-82-4
Molekylær formel C7H7D6NO2
Mængde 25 mg
Synonym (E)-4-(Dimethylamino)but-2-enoic Acid-d6 Methyl Ester, (E)-4-(Dimethylamino)-2-butenoic Acid-d6 Methyl Ester
InChI formel InChI=1S/C7H13NO2/c1-8(2)6-4-5-7(9)10-3/h4-5H,6H2,1-3H3/b5-4+/i1D3,2D3
SMIL [2H]C([2H])([2H])N(C\C=C\C(=O)OC)C([2H])([2H])[2H]
IUPAC navn methyl (E)-4-[bis(trideuteriomethyl)amino]but-2-enoate
Molekylvægt (g/mol) 149.22
Formel vægt 149.1323