Få mere at vide
trans-3'-Hydroxy Cotinine-d3 (Racemic Mixtures), TRC
Artikelnummer. 30403994
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30403994 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403994 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9245101MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-3'-Hydroxycotinine-D3
CAS 159956-78-2
Renhedsgrad noter HPLC
Molekylær formel C10 2H3 H9 N2 O2
Mængde 1 mg
Synonym 2-Pyrrolidinone, 3-hydroxy-1-(methyl-d3)-5-(3-pyridinyl)-, trans- (9CI), 2-Pyrrolidinone, 3-hydroxy-1-(methyl-d3)-5-(3-pyridinyl)-, trans-(±)-
InChI formel InChI=1S/C10H12N2O2/c1-12-8(5-9(13)10(12)14)7-3-2-4-11-6-7/h2-4,6,8-9,13H,5H2,1H3/t8-,9+/m0/s1/i1D3
SMIL [2H]C([2H])([2H])N1[C@@H](C[C@@H](O)C1=O)c2cccnc2
IUPAC navn (3R,5S)-3-hydroxy-5-pyridin-3-yl-1-(trideuteriomethyl)pyrrolidin-2-one
Molekylvægt (g/mol) 195.23
Formel vægt 195.1087
Procent renhed >95