Få mere at vide
trans-2-Methyl-3-(5'-carboxy-3'-methyl-2'-pentenyl)-1,4-naphthoquinone, TRC
Artikelnummer. 30479854
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479854 10 mg
30474917 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479854 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29408010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34185-71-2
Molekylær formel C18H18O4
Molekylvægt (g/mol) 298.33
Synonym (E)-6-(1,4-Dihydro-3-methyl-1,4-dioxo-2-naphthalenyl)-4-methyl-4-hexenoic Acid
SMIL O=C(C1=C2C=CC=C1)C(C/C=C(C)/CCC(O)=O)=C(C)C2=O
Tekniske data
Kemisk navn eller materiale trans-2-Methyl-3-(5'-carboxy-3'-methyl-2'-pentenyl)-1,4-naphthoquinone
Mængde 10 mg
InChI formel InChI=1S/C18H18O4/c1-11(8-10-16(19)20)7-9-13-12(2)17(21)14-5-3-4-6-15(14)18(13)22/h3-7H,8-10H2,1-2H3,(H,19,20)/b11-7+
Formel vægt 298.12