missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Hexen-1-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-7021080
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
928-95-0
C6H12O
100.16
MFCD00002927
ZCHHRLHTBGRGOT-SNAWJCMRSA-N
trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
5318042
CCC\C=C\CO
This item is not returnable. View return policy
928-95-0
C6H12O
100.16
MFCD00002927
ZCHHRLHTBGRGOT-SNAWJCMRSA-N
trans-2-hexen-1-ol, trans-2-hexenol, 2-hexenol, e-hex-2-en-1-ol, e-2-hexen-1-ol, 2-hexen-1-ol, 2-hexen-1-ol, 2e, 3-propylallyl alcohol, 2-hexen-1-ol, e, 2-hexen-1-ol, trans
5318042
CCC\C=C\CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.843
Boiling Point 158°C to 160°C
Flash Point 54°C (129°F)
Odor Pleasant
Refractive Index 1.438
Quantity 25 g
UN Number UN1987
Beilstein 1719709
IUPAC Name (2E)-hex-2-en-1-ol
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material trans-2-Hexen-1-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'