missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-Amino-1-cyclohexanecarboxylic acid, 98%, ACROS Organics™
1487.00 DKK - 4609.00 DKK
Chemical Identifiers
CAS | 5691-19-0 |
---|---|
Molecular Formula | C7H13NO2 |
Molecular Weight (g/mol) | 143.19 |
MDL Number | MFCD00145418 |
InChI Key | USQHEVWOPJDAAX-PHDIDXHHSA-N |
Synonym | 1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid # |
PubChem CID | 2724640 |
IUPAC Name | (1R,2R)-2-aminocyclohexane-1-carboxylic acid |
SMILES | C1CCC(C(C1)C(=O)O)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10412061
|
Acros Organics
265630010 |
1g | Glass bottle |
1487.00 DKK
1g |
In stock, ready to ship: 1 | Please sign in to purchase this item. Need a web account? Register with us today! | |||
10016343
|
Acros Organics
265630050 |
5g | Glass bottle |
4609.00 DKK
5g |
Estimated Shipment 12-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Chemical Identifiers
5691-19-0 | |
143.19 | |
USQHEVWOPJDAAX-PHDIDXHHSA-N | |
2724640 | |
C1CCC(C(C1)C(=O)O)N |
C7H13NO2 | |
MFCD00145418 | |
1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid # | |
(1R,2R)-2-aminocyclohexane-1-carboxylic acid |
Specifications
trans-2-Amino-1-cyclohexanecarboxylic acid | |
Glass bottle | |
274°C to 278°C | |
5691-19-0 | |
C7H13NO2 | |
1r,2r-2-aminocyclohexanecarboxylic acid, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid, 1r,2r-2-aminocyclohexane-1-carboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 2-amino-, trans, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, 2-nchca, 2-aminocyclohexanecarboxylic acid # | |
USQHEVWOPJDAAX-PHDIDXHHSA-N | |
(1R,2R)-2-aminocyclohexane-1-carboxylic acid | |
2724640 | |
Powder and Chunks |
Authentic | |
White | |
1g | |
98% | |
MFCD00145418 | |
Solubility in water: soluble in water. Other solubilities: soluble in methanol | |
C1CCC(C(C1)C(=O)O)N | |
143.19 | |
143.19 | |
98% |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- trans-2-Amino-1-cyclohexanecarboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.