Få mere at vide
trans-1-Cinnamylpiperazine Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | trans-1-Cinnamylpiperazine Dihydrochloride |
| CAS | 163596-56-3 |
| Molekylær formel | C13H18N2 . 2(HCl) |
| Mængde | 10 g |
| Synonym | (E)-1-(3-Phenyl-2-propenyl)piperazine Dihydrochloride, 1-[(2E)-3-Phenyl-2-propenyl]piperazine Dihydrochloride, (E)-N-Cinnamylpiperazine Dihydrochloride, 1-(trans-Cinnamyl)piperazine Dihydrochloride, 1-[(2E)-3-Phenylprop-2-enyl]piperazine Dihydrochloride, 1-[(E)-1-Phenyl-1-propen-3-yl]piperazine Dihydrochloride, 1-[(E)-3-Phenylprop-2-enyl]piperazine Dihydrochloride, 1-trans-1-Cinnamylpiperazine Dihydrochloride, N-((E)-Cinnamyl)piperazine Dihydrochloride |
| InChI formel | InChI=1S/C13H18N2.ClH/c1-2-5-13(6-3-1)7-4-10-15-11-8-14-9-12-15;/h1-7,14H,8-12H2;1H/b7-4+; |
| SMIL | Cl.Cl.C(\C=C\c1ccccc1)N2CCNCC2 |
| IUPAC navn | 1-[(E)-3-phenylprop-2-enyl]piperazine;hydrochloride |
| Molekylvægt (g/mol) | 238.75 |