Få mere at vide
trans 1-Benzyl-4-(4-fluorophenyl)-3-piperidinemethanol, TRC
Artikelnummer. 30345150
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30345150 50 mg
30341545 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30345150 Leverandør Toronto Research Chemicals Leverandørnr. TRCB27667050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans 1-Benzyl-4-(4-fluorophenyl)-3-piperidinemethanol
CAS 188869-25-2
Molekylær formel C19 H22 F N O
Mængde 50 mg
Synonym 3-Piperidinemethanol, 4-(4-fluorophenyl)-1-(phenylmethyl)-, (3R,4S)-rel-, 3-Piperidinemethanol, 4-(4-fluorophenyl)-1-(phenylmethyl)-, trans-, rel-(3R,4S)-4-(4-Fluorophenyl)-1-(phenylmethyl)-3-piperidinemethanol, trans-1-Benzyl-4-(4-fluorophenyl)-3-hydroxymethylpiperidine, trans-1-Benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol
InChI formel InChI=1S/C19H22FNO/c20-18-8-6-16(7-9-18)19-10-11-21(13-17(19)14-22)12-15-4-2-1-3-5-15/h1-9,17,19,22H,10-14H2/t17-,19-/m0/s1
SMIL OC[C@@H]1CN(Cc2ccccc2)CC[C@H]1c3ccc(F)cc3
IUPAC navn [(3S,4R)-1-benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol
Molekylvægt (g/mol) 299.38
Formel vægt 299.1685