missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, 98%, Thermo Scientific Chemicals

Catalog Number p-7026742
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
125572-95-4
C14H20N2O8
344.32
MFCD00149243,MFCD00066429,MFCD00003845
FCKYPQBAHLOOJQ-NXEZZACHSA-L
2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
2723844
2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O
This item is not returnable. View return policy
125572-95-4
C14H20N2O8
344.32
MFCD00149243,MFCD00066429,MFCD00003845
FCKYPQBAHLOOJQ-NXEZZACHSA-L
2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
2723844
2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate is an analytical reagent and a complexating agent for sodium. It acts as a ligand to prepare lanthanide shift reagents. It is involved in the separation and determination of iron(III)-dimethyldithiocarbamate, manganese(II)-ethylenebisdithiocarbamate and zinc(II)-ethylenebisdithiocarbamate by sensitive capillary electrophoretic ultraviolet method.

Solubility
Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼220°C (decomposition)
Quantity 25 g
Beilstein 2226756
Solubility Information Slightly soluble in water. Soluble in 1N sodium hydroxide and alkali solutions. Insoluble in most common organic solvents.
Formula Weight 364.35 (346.34 Anhydrous)
Percent Purity 98%
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
compliance-icons
Product Identifier
  • trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'