missing translation for 'onlineSavingsMsg'
Learn More

Trandolaprilat Methyl Ester, TRC

Product Code. 30452711
Change view
Click to view available options
Mængde:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30452711 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452711 Supplier Toronto Research Chemicals Supplier No. TRCT7135302MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2S,3aR,7aS)-1-[N-[(S)-1-carboxy-3-phenylpropyl]-L-alanyl]hexahydro-2-indolinecarboxylic acid 1-methyl ester
CAS 118194-41-5
Molekylær formel C23 H32 N2 O5
Mængde 2 mg
Synonym Trandolapril Related Compound A
InChI formel InChI=1S/C23H32N2O5/c1-15(24-18(23(29)30-2)13-12-16-8-4-3-5-9-16)21(26)25-19-11-7-6-10-17(19)14-20(25)22(27)28/h3-5,8-9,15,17-20,24H,6-7,10-14H2,1-2H3,(H,27,28)/t15-,17+,18-,19-,20-/m0/s1
SMIL COC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2[C@H]3CCCC[C@@H]3C[C@H]2C(=O)O
IUPAC navn (2S,3aR,7aS)-1-[(2S)-2-[[(2S)-1-methoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid
Molekylvægt (g/mol) 416.51
Formel vægt 416.2311
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.