Learn More
Thermo Scientific™ TRACE™ TR-1 GC Columns
Use this non-polar GC column for method development and analysis of chlorinated and nitroaromatic compounds and environmental samples.
Brand:  Thermo Scientific™ 260A470P
5465.15 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10311545
  • 5492.48 DKK / Each
Estimated Shipment: 16-06-2025
Explore more special offers
Description
Use the Thermo Scientific™ TRACE™ TR-1 GC column for method development and chlorinated and nitroaromatic compounds and environmental analyses. These non-polar phase, 100% dimethyl polysiloxane gas chromatography columns have a high operating temperature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.53 mm
TR-1
30 m
TRACE
5 μm
Product Title
Thermo Scientific™ TRACE™ TR-1 GC Columns > 30m x 0.53mm x 5μm