Få mere at vide
Trace Metals III for AA and ICP-OES, 6 Components, Honeywell Fluka™
Shop alle Honeywell Fluka produkter
Mængde:
100 ml
Beskrivelse
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Trace Metals III for AA and ICP-OES |
| Tæthed | 1.06 g/cm3 |
| pH | <2 |
| Mængde | 100 ml |
| Grad | Standards for ICP and ICP-MS |