Få mere at vide
Trace Metals II for AA and ICP-OES, 3 Components, Honeywell Fluka™
Artikelnummer. 16606093 Shop alle Honeywell Fluka produkter
Mængde:
100 ml
Artikelnummer. 16606093
Brand: Honeywell Fluka E2167100ML
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Trace Metals II for AA and ICP-OES
Tæthed 1.05 g/cm3
pH <2
Mængde 100 ml
Grad Standards for ICP and ICP-MS