Få mere at vide
Ansell™ TouchNTuff™ 69-210 Series Natural Rubber Latex Gloves
Shop alle Ansell produkter
Størrelse:
Small
Medium
Large
Pakningsstørrelse:
Pakke med 100
Beskrivelse
- Combined safety and comfort for delicate handling.
- Ambidextrous, same glove fits both hands
- Convenient and economical - stocks can be kept to a minimum
- Flexible and comfortable to wear - cooler than vinyl, with superior finger flexibility and grip
- Available in both powdered (69-210) and powder-free (69-318) versions
Tekniske data
Tekniske data
| Certificeringer/compliance | EN 420:2003 + A1:2009 and EN 374:2003 |
| Farve | Natural |
| Latex fri | No |
| Materiale | Rubber Latex |
| Pulveriseret | Powdered |
| Produkttype | Natural Rubber Latex Gloves |
| Størrelse | Medium |
| Sterilitet | Non-sterile |
| Tykkelse (metrisk) | 0.10 mm |
| Emballagetype | 100 gloves per box, 10 boxes per carton/case |
| Vis mere |