Learn More
Amarell™ Total Immersion Laboratory Thermometers
Brand: Amarell™ L33038
Additional Details : Weight : 0.02400kg
Specifications
DIN | |
300 mm | |
Thermometer | |
-50°C | |
5°C |
Total Immersion | |
Toluene | |
50°C | |
-50°C to +50°C |