Få mere at vide
Tosylsarcosine-d3, TRC
Artikelnummer. 30454929
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30454929 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454929 Leverandør Toronto Research Chemicals Leverandørnr. TRCT68626125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tosylsarcosine-d3
CAS 35009-46-2
Molekylær formel C10H10D3NO4S
Mængde 25 mg
Synonym N-(Methyl-d3)-N-[(4-methylphenyl)sulfonyl]-glycine, N-(p-Tolylsulfonyl)-sarcosine-d3, N-Methyl-d3-N-[(4-methylphenyl)sulfonyl]glycine, N-Tosylsarcosine-d3
InChI formel InChI=1S/C10H13NO4S/c1-8-3-5-9(6-4-8)16(14,15)11(2)7-10(12)13/h3-6H,7H2,1-2H3,(H,12,13)/i2D3
SMIL [2H]C([2H])([2H])N(CC(=O)O)S(=O)(=O)c1ccc(C)cc1
IUPAC navn 2-[(4-methylphenyl)sulfonyl-(trideuteriomethyl)amino]acetic acid
Molekylvægt (g/mol) 246.3
Formel vægt 246.0754