missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ TORC2 Antibody Blocking Peptide
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Mængde:
0,1 mg
Tekniske data
Tekniske data
| Gen-id (Entrez) | 200186 |
| Koncentration | 1 mg/mL |
| Indhold og opbevaring | Store at -80°C. Avoid freeze-thaw cycles. |
| Til brug med (applikation) | This peptide is useful as a blocking peptide for NBP2-22356. For further blocking peptide related protocol, click here . |
| Gene Alias | CREB regulated transcription coactivator 2, CREB-regulated transcription coactivator 2, TORC-2, TORC2transducer of regulated cAMP response element-binding protein (CREB) 2, Transducer of CREB protein 2, Transducer of regulated cAMP response element-binding protein 2 |
| Gen symbol | CRTC2 |
| Mængde | 0,1 mg |
| Forskningskategori | Cancer, Hypoxia, Lipid and Metabolism, Signal Transduction, Transcription Factors and Regulators |
| Opbevaringsbuffer | Peptide dissolved in dH20. Contains no BSA. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion