Få mere at vide
Torasemide, Anhydrous, LoGiCal
Artikelnummer. 30458171
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458171 10 mg
1 options
Artikelnummer. 30458171 Leverandør LoGiCal Leverandørnr. LGCFOR1283.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Torasemide
CAS 56211-40-6
Analyt- eller komponentnavne Torasemide
Forsendelsestilstand Room Temperature
Molekylær formel C16 H20 N4 O3 S
Certificeringer/compliance ISO 9001
Synonym Torasemide anhydrous, 1-(1-Methylethyl)-3-[[4-[(3-methylphenyl)amino]pyridin-3-yl]sulfonyl]urea, N-[[(1-Methylethyl)amino]carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide
InChI formel InChI=1S/C16H20N4O3S/c1-11(2)18-16(21)20-24(22,23)15-10-17-8-7-14(15)19-13-6-4-5-12(3)9-13/h4-11H,1-3H3,(H,17,19)(H2,18,20,21)
Løsningstype Neat
SMIL CC(C)NC(=O)NS(=O)(=O)c1cnccc1Nc2cccc(C)c2
IUPAC navn 1-[4-(3-methylanilino)pyridin-3-yl]sulfonyl-3-propan-2-ylurea
Industry Type Clinical Testing
Molekylvægt (g/mol) 348.42
Mængde 10 mg
Formel vægt 348.1256 g/mol