Learn More
DWK Life Sciences DURAN™ Weighing Bottle Tops, unground
6265.25 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Description
DURAN™ weighing bottles are used for weighing chemicals on analysis trolleys. The tight seal of the replaceable, ground weighing glass lid prevents sample loss during transport after weighing.
- Unground
- Subsequent grinding results in a tight seal of the weighing lid in the weighing pan. A loss of sample can thus be avoided.
- Grinding glass product
Specifications
Closure Size | NS 24 |
For Use With (Application) | Weighing of chemicals |
Material | Borosilicate Glass (3.3) |
Quantity | 240/Pk. |
Autoclavable | Autoclavable |