Learn More
DWK Life Sciences DURAN™ Weighing Bottle Tops, unground
Catalog Number 15854848
6265.25 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Weighing glass lid for grinding and completing.
DURAN™ weighing bottles are used for weighing chemicals on analysis trolleys. The tight seal of the replaceable, ground weighing glass lid prevents sample loss during transport after weighing.
  • Unground
  • Subsequent grinding results in a tight seal of the weighing lid in the weighing pan. A loss of sample can thus be avoided.
  • Grinding glass product
Closure Size NS 24
For Use With (Application) Weighing of chemicals
Material Borosilicate Glass (3.3)
Quantity 240/Pk.
Autoclavable Autoclavable