Få mere at vide
Topotecan-d5, TRC
Artikelnummer. 30453880
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30453880 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453880 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5425031MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Topotecan-d5
CAS 1133355-98-2
Molekylær formel C23H18D5N3O5
Mængde 1 mg
Synonym (4S)-10-[(Dimethylamino)methyl]-4-(ethyl-d5)-4,9-dihydroxy-1H-pyrano[3’,4’,6-7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, NSC-609669-d5, SKF-104864A-d5, Hycamtin-d5
InChI formel InChI=1S/C23H23N3O5/c1-4-23(30)16-8-18-20-12(9-26(18)21(28)15(16)11-31-22(23)29)7-13-14(10-25(2)3)19(27)6-5-17(13)24-20/h5-8,27,30H,4,9-11H2,1-3H3/t23-/m0/s1/i1D3,4D2
SMIL [2H]C([2H])([2H])C([2H])([2H])[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(CN(C)C)c(O)ccc5nc34)C2=O
Molekylvægt (g/mol) 426.48
Formel vægt 426.1952