missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Topoisomerase II alpha Mouse, Clone: TOP2A/1362, enQuireBio™
Mouse Monoclonal Antibody
1480.00 DKK - 3660.00 DKK
Specifications
Antigen | Topoisomerase II alpha |
---|---|
Clone | TOP2A/1362 |
Concentration | 200ÎĽg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960998
|
EnquireBio
7153-MSM2-P0 |
20 ÎĽg | N/A | ||||||
15970998
|
EnquireBio
7153-MSM2-P1 |
100 ÎĽg |
3660.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Topoisomerase II alpha | |
200ÎĽg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
7153 | |
Recombinant human Topoisomerase II alpha fragment (aa1352-1493) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
170kDa |
TOP2A/1362 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
XP_011523467 | |
TOP2A | |
IgG2b Îş | |
Protein A/G Purified | |
Nuclear | |
HeLa cells or Tonsil |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Topoisomerase II alpha Mouse, Clone: TOP2A/1362, enQuireBio™