Learn More
Invitrogen™ TOPO™ XL-2 Complete PCR Cloning Kit, with One Shot™ OmniMAX™ 2 T1R Chemically Competent E. coli Cells
Product Code. 15867894
20 Reactions, 20 reactions
Click to view available options
No. of Reactions:
10 Reactions
20 Reactions
Unit Size:
20 reactions
Each
Este artĂ­culo no se puede devolver. Vea la polĂ­tica de devoluciones
For Research Use Only. Not for use in diagnostic procedures.
Este artĂ­culo no se puede devolver. Vea la polĂ­tica de devoluciones
For Research Use Only. Not for use in diagnostic procedures.
Includes
Box 1:
  • pCR XL-2-TOPO vector
  • Salt Solution
  • T3 Primer
  • T7 Primer
  • XL Control PCR Template
  • XL Control PCR Primers
  • Platinum SuperFi Green PCR Master Mix
  • SuperFi GC Enhancer
  • Nuclease-free water
Box 2:
  • Gel Solubilization Buffer (L3)
  • Binding Buffer (B2)
  • Wash Buffer (W1)
  • Elution Buffer (E1)
  • PureLink Clean-up Spin Columns (in Wash Tubes)
  • PureLink Recovery/Elution Tubes
Box 3:
  • One Shot OmniMAX 2 T1R chemically competent E. coli
  • S.O.C. Medium
  • pUC 19 Control DNA
Provides all the necessary elements for efficient cloning of extra-long PCR products (up to 13 kb)
The kit uses the linearized and topoisomerase 1-activated pCR™-XL-2-TOPO™ vector, which is compatible with the cloning of blunt-end PCR fragments. Amplification of long PCR fragments is enabled by Platinum™SuperFi™ Green PCR Master Mix, which is included in the kit. Topoisomerase I activation of the vector allows PCR products to be ligated in just 5 minutes on your bench–top, resulting in high cloning efficiency (up to 90% positive recombinants with 10 kb fragments).
The pCR-XL-2-TOPO vector includes:
  • ccdB gene for positive selection
  • EcoR I site flanking the PCR product insertion site for easy excision of inserts
  • Ampicillin- and kanamycin-resistance genes for your choice of selection in E. coli
  • T7 promoter/priming site for in vitro RNA transcription and sequencing
  • T7, T3, and M13 forward and reverse primer sites for sequencing
The TOPO XL-2 Complete PCR Cloning Kit includes:
  • TOPO XL-2 PCR Cloning Kits containing the pCR-XL-2-TOPO vector
  • Platinum SuperFi Green PCR Master Mix—featuring a proofreading DNA polymerase (>100X fidelity compared to Taq) with high processivity and a density gradient to generate accurate, long PCR amplicons ready to load onto an agarose gel for gel extraction
  • PureLink™ Quick Gel Extraction and PCR Purification Combo Kit—designed to purify DNA fragments from agarose gels or direct PCR purification using a silica-based spin cartridge in less than 30 minutes
  • One Shot™ OmniMAX™ 2 T1R Chemically Competent E. coli Cells—an improved chemically competent cell line, perfect for use in all cloning applications.
Especificaciones
Bacterial or Yeast Strain OmniMAX2
Antibiotic Resistance Bacterial Ampicillin (AmpR), Kanamycin (KanR)
Cell Type Chemically Competent E. coli
Cloning Method Zero Blunt TOPO
Content And Storage Box 1: Store at -5 to -30°C.
• pCR XL-2-TOPO vector
• Salt Solution
• T3 Primer
• T7 Primer
• XL Control PCR Template
• XL Control PCR Primers
• Platinum SuperFi Green PCR Master Mix
• SuperFi GC Enhancer
• Nuclease-free water
Box 2: Store at room temperature.
• Gel Solubilization Buffer (L3)
• Binding Buffer (B2)
• Wash Buffer (W1)
• Elution Buffer (E1)
• PureLink Clean-up Spin Columns (in Wash Tubes)
• PureLink Recovery/Elution Tubes
Box 3:
• One Shot OmniMAX 2 T1R chemically competent E. coli
• S.O.C. Medium
• pUC 19 Control DNA
Store competent cells and pUC19 at -65 to -85°C.
Format Kit
For Use With (Application) PCR Cloning
No. of Reactions 20 Reactions
Product Line TOPO™
Product Type Cloning Kit
Promoter T3, T7
Quantity 20 reactions
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Vector pCR-XL-2-TOPO
Mostrar más Mostrar menos
For Research Use Only. Not for use in diagnostic procedures.
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂ­tulo del producto
Invitrogen™ TOPO™ XL-2 Complete PCR Cloning Kit, with One Shot™ OmniMAX™ 2 T1R Chemically Competent E. coli Cells >
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado