Få mere at vide
Topiramate-d12, TRC
Artikelnummer. 30452831
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30452831 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452831 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5402522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Topiramate-D12
CAS 1279037-95-4
Renhedsgrad noter HPLC
Molekylær formel C12 2H12 H9 N O8 S
Mængde 2.5 mg
Synonym Topiramate-D12
InChI formel InChI=1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1/i1D3,2D3,3D3,4D3
SMIL [2H]C([2H])([2H])C1(O[C@@H]2CO[C@@]3(COS(=O)(=O)N)OC(O[C@H]3[C@@H]2O1)(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn [(3aS,5aR,8aR,8bS)-2,2,7,7-tetrakis(trideuteriomethyl)-5,5a,8a,8b-tetrahydrodi[1,3]dioxolo[5,4-b:5',4'-d]pyran-3a-yl]methyl sulfamate
Molekylvægt (g/mol) 351.44
Formel vægt 351.1741
Procent renhed >95