Få mere at vide
Topiramate-d12 (di-iso-propylidene-d12), CDN
Artikelnummer. 30319603
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319603 0.01 g
30318465 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319603 Leverandør CDN Leverandørnr. CDND69060.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Topiramate-D12
CAS 1279037-95-4
Molekylær formel C12 2H12 H9 N O8 S
Mængde 0.01 g
Synonym Topiramate-D12
InChI formel InChI=1 S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9 H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1/i1D3,2D3,3D3,4D3
SMIL [2 H]C([2 H])([2 H])C1(O[C@@H]2 CO[C@@]3(COS(=O)(=O)N)OC(O[C@H]3 [C@@H]2O1)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn [(3 AS,5 aR,8 aR,8 bS)-2,2,7,7-tetrakis(trideuteriomethyl)-5,5 A,8 A,8 b-tetrahydrodi[1,3 ]dioxolo[5,4-b:5',4'-d]pyran-3 A-yl]methyl sulfamate
Molekylvægt (g/mol) 351.44
Formel vægt 351.1741 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity