Learn More
Top Loading Kit, for OHAUS™ Scout STX and SKX Precision Portable Balances
For use with Scout STX and SKX series Precision Portable Balances.
Brand:  Ohaus 30269020
Additional Details : Weight : 0.50000kg
Product Code. 15569641
  • 170.00 DKK / Each
Estimated Shipment: 12-06-2024
to see stock.
Description
Description
Allows balance to be safely stored away when not in use.
balance, density, solids, liquid
Specifications
Specifications
Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Density Determination Kits