Learn More
BINDER™ Top Access Port (10 to 50 mm)
For Use With: FD 23, FD 56, FD 115, FD 260, FD 720
Brand: BINDER™ 8012-1380
4506.42 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use on top
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 50 mm, top | |
Steel | |
Silicone plug |
BINDER Chambers Series FD | |
Access Port | |
50 mm |
Product Content Correction
Product Title
BINDER™ Top Access Port (10 to 50 mm) > Diameter: 50 mm, For Use With: FD 23, FD 56, FD 115, FD 260, FD 720
Spot an opportunity for improvement?Share a Content Correction