Få mere at vide
(±)-Tonalide-d3 (acetyl-d3), CDN
Artikelnummer. 30313544
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313544 0.1 g
30314786 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313544 Leverandør CDN Leverandørnr. CDND81000.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale AHTN (Tonalide) (6-acetyl) D3
CAS 1396967-82-0
Molekylær formel C18 2H3 H23 O
Mængde 0.1 g
InChI formel InChI=1 S/C18H26O/c1-11-8-16-15(9-14(11)13(3)19)17(4,5)10-12(2)18(16,6)7/h8-9,12 H,10H2,1-7H3/i3D3
SMIL [2 H]C([2 H])([2 H])C(=O)c1cc2c(cc1C)C(C)(C)C(C)CC2(C)C
IUPAC navn 2,2,2-trideuterio-1-(3,5,5,6,8,8-hexamethyl-6,7-dihydronaphthalen-2-yl)ethanone
Molekylvægt (g/mol) 261.42
Formel vægt 261.2172 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity