missing translation for 'onlineSavingsMsg'
Learn More

Tomatidine Hydrochloride ( >85%), TRC

Product Code. 30454868
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30454868 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454868 Supplier Toronto Research Chemicals Supplier No. TRCT53850010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Tomatidine hydrochloride
CAS 6192-62-7
Molekylær formel C27 H45 NO2 . x HCl
Mængde 10 mg
Synonym (3beta,5alpha,22beta,25S)-Spirosolan-3-ol Hydrochloride, Tomatidin Hydrochloride
InChI formel InChI=1S/C27H45NO2.ClH/c1-16-7-12-27(28-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(29)8-10-25(18,3)21(20)9-11-26(22,24)4;/h16-24,28-29H,5-15H2,1-4H3;1H/t16-,17-,18-,19-,20+,21-,22-,23-,24-,25-,26-,27+;/m0./s1
SMIL O[C@H]1CC[C@@]2(C)[C@](C1)(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])C[C@@]5([H])[C@]4([H])[C@H](C)[C@@]6(NC[C@@H](C)CC6)O5)[H].[xHCl]
Molekylvægt (g/mol) 452.11
Formel vægt 451.3217
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.