Få mere at vide
Tolvaptan-d7, TRC
Artikelnummer. 30450295
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30450295 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30450295 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53665225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolvaptan-d7
CAS 1246818-18-7
Renhedsgrad noter HPLC
Molekylær formel C26H18D7ClN2O3
Mængde 25 mg
Synonym N-[4-[(7-Chloro-2,3,4,5-tetrahydro-5-hydroxy-1H-1-benzazepin-1-yl)carbonyl]-3-methylphenyl]-2-methylbenzamide-d7, OPC 41061-d7
InChI formel InChI=1S/C26H25ClN2O3/c1-16-6-3-4-7-20(16)25(31)28-19-10-11-21(17(2)14-19)26(32)29-13-5-8-24(30)22-15-18(27)9-12-23(22)29/h3-4,6-7,9-12,14-15,24,30H,5,8,13H2,1-2H3,(H,28,31)/i1D3,3D,4D,6D,7D
SMIL O=C(NC1=CC=C(C(N2CCCC(O)C3=C2C=CC(Cl)=C3)=O)C(C)=C1)C4=C([2H])C([2H])=C([2H])C([2H])=C4C([2H])([2H])[2H]
Molekylvægt (g/mol) 455.98
Formel vægt 455.1993
Procent renhed >95