missing translation for 'onlineSavingsMsg'
Få mere at vide

Toluidine Red, TRC

Artikelnummer. 30137713
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137713 10 g
30137068 25 g
17967154 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30137713 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53622010G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2425-85-6
Molekylær formel C17 H13 N3 O3
Molekylvægt (g/mol) 307.3
Synonym 2-Naphthalenol, 1-[2-(4-methyl-2-nitrophenyl)diazenyl]- (ACI), 1-[2-(4-Methyl-2-nitrophenyl)diazenyl]-2-naphthalenol (ACI), 2-Naphthalenol, 1-[(4-methyl-2-nitrophenyl)azo]- (9CI), C.I. Pigment Red 3 (7CI, 8CI), Toluidine red (6CI), 1-((4-Methyl-2-nitrophenyl)diazenyl)naphthalen-2-ol, 1-(4-Methyl-2-nitrophenylazo)-2-naphthol, 1-(o-Nitro-p-tolylazo)-2-naphthol, 1-[(2-Nitro-4-methylphenyl)azo]-2-naphthol, 2-Nitro-p-toluidine-2-naphthol, Accosperse Toluidine Red XL, ADC Toluidine Red B, Atlasol Spirit Red 3, C.I. 12120, C.P. Toluidine Toner A 2989, C.P. Toluidine Toner A 2990, C.P. Toluidine Toner Dark RS 3340, C.P. Toluidine Toner Deep X 1865, C.P. Toluidine Toner Light RS 3140, C.P. Toluidine Toner RT 6101, C.P. Toluidine Toner RT 6104, Calcotone Toluidine Red YP, Carnelio Helio Red, Chromatex Red J, D and C Red No. 35, Dainichi Permanent Red 4R, Deep Fastona Red, Duplex Toluidine Red L 20-3140, Eljon Fast Scarlet PV Extra, Eljon Fast Scarlet RN, Enialit Light Red RL, Fast Red A, Fast Red A (pigment), Fast Red AB, Fast Red J, Fast Red JE, Fast Red R, Fastona Red B, Fastona Scarlet RL, Fastona Scarlet YS, Graphtol Red A 4RL, Hansa Red B, Hansa Red G, Hansa Scarlet RB, Hansa Scarlet RN, Hansa Scarlet RNC, Helio Fast Red BN, Helio Fast Red RL, Helio Fast Red RN, Helio Red RL, Helio Red Toner, Hispalit Fast Scarlet RN, Independence Red, Irgalite Fast Red P 4R, Irgalite Fast Scarlet RND, Irgalite Red PV 2, Irgalite Red RNPX, Irgalite Scarlet RB, Isol Fast Red HB, Isol Fast Red RN 2B, Isol Fast Red RN 2G, Isol Fast Red RNB, Isol Fast Red RNG, Isol Toluidine Red HB, Isol Toluidine Red RN 2B, Isol Toluidine Red RN 2G, Isol Toluidine Red RNB, Isol Toluidine Red RNG, Japan Red 221, Japan Red No. 221, Kromon Helio Fast Red, Kromon Helio Fast Red YS, Lake Red 4R, Lake Red 4RII, Lithol Fast Scarlet RN, Lutetia Fast Red 3R, Lutetia Fast Scarlet RF, Lutetia Fast Scarlet RJN, Monolite Fast Scarlet CA, Monolite Fast Scarlet GSA, Monolite Fast Scarlet RB, Monolite Fast Scarlet RBA, Monolite Fast Scarlet RN, Monolite Fast Scarlet RNA, Monolite Fast Scarlet RNV, Monolite Fast Scarlet RT, No. 2 Forthfast Scarlet, NSC 45193, Oralith Red P 4R, Permanent Red 4R, Pigment Red 3, Pigment Red RL, Pigment Scarlet, Pigment Scarlet (Russian), Pigment Scarlet B, Pigment Scarlet Conc., Pigment Scarlet N, Pigment Scarlet R, Polymo Red FGN, Recolite Fast Red RBL, Recolite Fast Red RL, Recolite Fast Red RYL, Red No. 221, Sanyo Scarlet Pure, Sanyo Scarlet Pure No. 1000, Scarlet Pigment RN, Segnale Light Red 2B, Segnale Light Red B, Segnale Light Red BR, Segnale Light Red C 4R, Segnale Light Red RL, Seikafast Red 4R4016, Sico Red L 3750, Siegle Red 1, Siegle Red B, Siegle Red BB, Silogomma Red RLL, Silosol Red RBN, Silosol Red RN, Siloton Red BRLL, Siloton Red RLL, Symuler Fast Red 4R100, Symuler Fast Scarlet 4R, Syton Fast Scarlet RB, Syton Fast Scarlet RD, Syton Fast Scarlet RN, Tertropigment Red HAB, Tertropigment Scarlet LRN, Toluidene Red, Toluidine Red 10451, Toluidine Red 3B, Toluidine Red 4R, Toluidine Red B, Toluidine Red BFB, Toluidine Red BFGG, Toluidine Red D 28-3930, Toluidine Red Light, Toluidine Red M 20-3785, Toluidine Red R, Toluidine Red RB 0336C, Toluidine Red RT 115, Toluidine Red Toner, Toluidine Red XL 20-3050, Toluidine Toner, Toluidine Toner Dark 5040, Toluidine Toner HR-X 2700, Toluidine Toner HR-X 2741, Toluidine Toner Keep HR-X 2742, Toluidine Toner L 20-3300, Toluidine Toner RT 252, Versal Scarlet PRNL, Versal Scarlet RNL, Vulcafor Scarlet A
IUPAC navn 1-[(E)-(4-methyl-2-nitrophenyl)diazenyl]naphthalen-2-ol
SMIL Cc1ccc(N=Nc2c(O)ccc3ccccc23)c(c1)[N+](=O)[O-]

Tekniske data

Kemisk navn eller materiale Toluidine Red
Mængde 10 g
InChI formel InChI=1S/C17H13N3O3/c1-11-6-8-14(15(10-11)20(22)23)18-19-17-13-5-3-2-4-12(13)7-9-16(17)21/h2-10,21H,1H3
Formel vægt 307.0957
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.