Learn More
Toluene-d8, for NMR, 99.5% atom D, with 0.03% TMS, Thermo Scientific Chemicals
Catalog Number p-3734306
Click to view available options
Quantity:
5 mL
25 mL
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2037-26-5
C7H8
100.19
MFCD00044638
YXFVVABEGXRONW-JGUCLWPXSA-N
toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
74861
1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
CC1=CC=CC=C1
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2037-26-5
C7H8
100.19
MFCD00044638
YXFVVABEGXRONW-JGUCLWPXSA-N
toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
74861
1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
CC1=CC=CC=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 0.9400g/mL
Deuterium (D) 99.4% min.
Flash Point 4°C
Packaging Glass Bottle
Refractive Index 1.494
Quantity 5 mL
Melting Point -84.0°C
Boiling Point 110.0°C
Linear Formula C6D5CD3
Beilstein 05, IV, 797
Formula Weight 100.21
Grade NMR
Chemical Name or Material Toluene-d8, 99.5 atom%D
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Toluene-d8
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.