missing translation for 'onlineSavingsMsg'
Learn More

Toluene-d8, 99.6%(Isotopic), Thermo Scientific Chemicals

Catalog Number p-7050321
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2037-26-5
C7H8
100.19
MFCD00044638
YXFVVABEGXRONW-JGUCLWPXSA-N
toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
74861
1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
CC1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2037-26-5
C7H8
100.19
MFCD00044638
YXFVVABEGXRONW-JGUCLWPXSA-N
toluene-d8, 2h8 toluene, perdeuteriotoluene, benzene-d5, methyl-d3, perdeuterotoluene, benzene-d5-, methyl-d3, 1,2,3,4,5-pentadeuterio-6-trideuteriomethyl benzene, toluene d8, toluene-d8, 99 atom % d, toluene-d8, 99.6 atom % d
74861
1,2,3,4,5-pentadeuterio-6-(trideuteriomethyl)benzene
CC1=CC=CC=C1

Toluene is mainly used as a precursor to benzene. Paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, and leather tanners. And also used as an octane booster in gasoline fuels used in internal combustion engines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 0.94g/mL
Flash Point 10° (50°F)
Odor Aromatic
Quantity 1 g
Melting Point -93°C
Boiling Point 111°C
UN Number UN1294
Formula Weight 100.19
Percent Purity 99.6%
Chemical Name or Material Toluene-d8, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'