Få mere at vide
Toluene-4-d1, CDN
Artikelnummer. 30323005
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323005 0.25 g -
30311658 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323005 Leverandør CDN Leverandørnr. CDND77840.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Toluene-4-D
CAS 4409-83-0
Molekylær formel C7 D H7
Mængde 0.25 g
Synonym Benzene-d, 4-methyl- (9 CI, ACI), 4-Methylbenzene-d (ACI), Toluene-p-d (7 CI, 8 CI), p-Deuteriotoluene, Toluene-4-d, Toluene-4-D
InChI formel InChI=1 S/C7H8/c1-7-5-3-2-4-6-7/h2-6 H,1H3/i2D
SMIL [2 H]c1ccc(C)cc1
IUPAC navn 1-deuterio-4-methylbenzene
Molekylvægt (g/mol) 93.145
Formel vægt 93.069 g/mol
Procent renhed 96 atom % D, min 98% Chemical Purity