Få mere at vide
Tolrestat, TRC
Artikelnummer. 30138216
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138216 100 mg
30006327 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138216 Leverandør Toronto Research Chemicals Leverandørnr. TRCT535490100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 82964-04-3
Molekylær formel C16 H14 F3 N O3 S
Molekylvægt (g/mol) 357.35
Synonym Glycine, N-[[6-methoxy-5-(trifluoromethyl)-1-naphthalenyl]thioxomethyl]-N-methyl-, N-[[6-Methoxy-5-(trifluoromethyl)-1-naphthalenyl]thioxomethyl]-N-methylglycine, AY 27773, Alredase, Lorestat, Tolrestat, Tolrestatin
IUPAC navn 2-[[6-methoxy-5-(trifluoromethyl)naphthalene-1-carbothioyl]-methylamino]acetic acid
SMIL COc1ccc2c(cccc2c1C(F)(F)F)C(=S)N(C)CC(=O)O
Tekniske data
Kemisk navn eller materiale Tolrestat
Mængde 100 mg
InChI formel InChI=1S/C16H14F3NO3S/c1-20(8-13(21)22)15(24)11-5-3-4-10-9(11)6-7-12(23-2)14(10)16(17,18)19/h3-7H,8H2,1-2H3,(H,21,22)
Formel vægt 357.0646