Få mere at vide
Tolperisone-d10 Hydrochloride, TRC
Artikelnummer. 30451040
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451040 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451040 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53547710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolperisone-d10','Hydrochloride
CAS 1185160-65-9
Molekylær formel C16 2H10 H13 N O . Cl H
Mængde 10 mg
Synonym 3-(2,2,3,3,4,4,5,5,6,6-Decadeuteriopiperidin-1-yl)-2-methyl-1-(4-methylphenyl)propan-1-one hydrochloride, Tolperisone-d10 hydrochloride, Tolperisone D10 hydrochloride (piperazinyl D10)
InChI formel InChI=1S/C16H23NO.ClH/c1-13-6-8-15(9-7-13)16(18)14(2)12-17-10-4-3-5-11-17;/h6-9,14H,3-5,10-12H2,1-2H3;1H/i3D2,4D2,5D2,10D2,11D2;
SMIL Cl.[2H]C1([2H])N(CC(C)C(=O)c2ccc(C)cc2)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn 3-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)-2-methyl-1-(4-methylphenyl)propan-1-one;hydrochloride
Molekylvægt (g/mol) 291.88
Formel vægt 291.2174