Få mere at vide
Tolnaftate, Thermo Scientific™
Antifungal agent
Brand: Thermo Scientific Alfa Aesar J61834.03
| Mængde | 1g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in chloroform at 50 mg/ml. Sparingly soluble in ethanol or methanol
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
Kemiske identifikatorer
| 2398-96-1 | |
| 307.411 | |
| FUSNMLFNXJSCDI-UHFFFAOYSA-N | |
| 5510 | |
| O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate |
| C19H17NOS | |
| MFCD00056611 | |
| Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid | |
| CHEBI:9620 | |
| CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 |
Tekniske data
| Tolnaftate | |
| White | |
| C19H17NOS | |
| 14,9519 | |
| Soluble in chloroform at 50 mg/ml; Sparingly soluble in ethanol or methanol | |
| CC1=CC(=CC=C1)N(C)C(=S)OC2=CC3=CC=CC=C3C=C2 | |
| 307.411 | |
| CHEBI:9620 |
| 2398-96-1 | |
| 1g | |
| MFCD00056611 | |
| Methyl-(3-methylphenyl)carbamothioic acid O-2-naphthyl ester; Carbamothioic acid | |
| FUSNMLFNXJSCDI-UHFFFAOYSA-N | |
| O-naphthalen-2-yl N-methyl-N-(3-methylphenyl)carbamothioate | |
| 5510 | |
| 307.41 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.