Få mere at vide
Tolfenamic-d4 Acid (benzoic-3,4,5,6-d4), CDN
Artikelnummer. 30321378
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321378 0.005 g
30318371 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321378 Leverandør CDN Leverandørnr. CDND77250.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolfenamic Acid-d4
CAS 1246820-82-5
Molekylær formel C14 D4 H8 Cl N O2
Mængde 0.005 g
Synonym 2-[(3-Chloro-2-methylphenyl)amino]-3,4,5,6-tetradeuteriobenzoic acid, Tolfenamic-d4 Acid (benzoic-3,4,5,6-d4), Tolfenamic Acid-d4, Tolfenamic Acid-D4 (benzoic-3,4,5,6-D4), 2-[(3-Chloro-2-methylphenyl)amino]benzoic-3,4,5,6-d4 acid, N-(2-Methyl-3-chlorophenyl)anthranilic acid-3,4,5,6-d4
InChI formel InChI=1 S/C14H12ClNO2/c1-9-11(15)6-4-8-12(9)16-13-7-3-2-5-10(13)14(17)18/h2-8,16 H,1H3,(H,17,18)/i2D,3 D,5 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(Nc2cccc(Cl)c2C)c1[2 H]
IUPAC navn 2-(3-chloro-2-methylanilino)-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 265.728
Formel vægt 265.081 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity