Få mere at vide
Tolfenamic Acid-d4, TRC
Artikelnummer. 30451609
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30451609 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451609 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5353025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolfenamic Acid-d4
CAS 1246820-82-5
Molekylær formel C14 D4 H8 Cl N O2
Mængde 5 mg
Synonym 2-[(3-Chloro-2-methylphenyl)amino]-3,4,5,6-tetradeuteriobenzoic acid, Tolfenamic-d4 Acid (benzoic-3,4,5,6-d4), Tolfenamic Acid-d4, Tolfenamic Acid-D4 (benzoic-3,4,5,6-D4), 2-[(3-Chloro-2-methylphenyl)amino]benzoic-3,4,5,6-d4 acid, N-(2-Methyl-3-chlorophenyl)anthranilic acid-3,4,5,6-d4
InChI formel InChI=1S/C14H12ClNO2/c1-9-11(15)6-4-8-12(9)16-13-7-3-2-5-10(13)14(17)18/h2-8,16H,1H3,(H,17,18)/i2D,3D,5D,7D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(Nc2cccc(Cl)c2C)c1[2H]
IUPAC navn 2-(3-chloro-2-methylanilino)-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 265.728
Formel vægt 265.081