Få mere at vide
Tolfenamic acid, 99+%, Thermo Scientific™
NSAID that activates calcium-activated potassium channels
Brand:  Thermo Scientific Alfa Aesar J61256.06
 Se flere versioner af dette produkt
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Produktkode 15463869
  • ONLINE EXCLUSIVE PRICE
    753.74 DKK
    677.65 DKK / 5g
    Spare 76.09 DKK
    10% Slukket
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble in water (slightly), acetone (∼10 mg/ml), DMSO (52 mg/ml at 25°C), methanol (∼10 mg/ml) and ethanol (50 mg/ml).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.