Få mere at vide
Tolclofos-methyl, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tolclofos-methyl |
| CAS | 57018-04-9 |
| Molekylær formel | C9 H11 Cl2 O3 P S |
| Mængde | 5 g |
| Synonym | Phosphorothioic acid','O-(2,6-dichloro-4-methylphenyl) O,O-dimethyl ester (9CI','ACI), O,O-Dimethyl O-(2,6-dichloro-4-methylphenyl) phosphorothioate, O-(2,6-Dichloro-4-methylphenyl) O,O-dimethyl phosphorothioate, Rhizolex, Rhizolex T 50, Rizolex, Tolclofos-methyl, Tolclophos-methyl |
| InChI formel | InChI=1S/C9H11Cl2O3PS/c1-6-4-7(10)9(8(11)5-6)14-15(16,12-2)13-3/h4-5H,1-3H3 |
| SMIL | COP(=S)(OC)Oc1c(Cl)cc(C)cc1Cl |
| IUPAC navn | (2,6-dichloro-4-methylphenoxy)-dimethoxy-sulfanylidene-λ^{5}-phosphane |
| Molekylvægt (g/mol) | 301.13 |
| Formel vægt | 299.9544 |