Få mere at vide
Tolcapone-d4, TRC
Artikelnummer. 30456421
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30456421 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30456421 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53525210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolcapone-d4
CAS 1246816-93-2
Molekylær formel C14H7D4NO5
Mængde 10 mg
Synonym (3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl-d4)methanone, Ro-40-7592-d4, Tasmar-d4
InChI formel InChI=1S/C14H11NO5/c1-8-2-4-9(5-3-8)13(17)10-6-11(15(19)20)14(18)12(16)7-10/h2-7,16,18H,1H3/i2D,3D,4D,5D
SMIL O=C(C1=C([2H])C([2H])=C(C)C([2H])=C1[2H])C2=CC(O)=C(O)C([N+]([O-])=O)=C2
Molekylvægt (g/mol) 277.27
Formel vægt 277.0888