Få mere at vide
Tocris Bioscience™ Tolcapone
GREENER_CHOICE
COMT inhibitor; also inhibits transthyretin aggregation
Brand:  Tocris Bioscience™ 5864/50
Encompass
Produktkode 15727376
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Tolcapone is a COMT inhibitor. Inhibits both brain and peripheral COMT. Also binds transthyretin (TTR) with high affinity (Kd1 and Kd2 values are 21 and 58 nM, respectively). Inhibits TTR aggregation in human plasma and prevents TTR-induced cytotoxicity in vitro. Stabilizes TTR in mice and humans in vivo. Orally bioavailable.
Tekniske data
Tekniske data
(3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl)methanone
134308-13-7
Tasmar, Ro 40-7592
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner