Få mere at vide
Tolbutamide-d9, TRC
Artikelnummer. 30454382
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30454382 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454382 Leverandør Toronto Research Chemicals Leverandørnr. TRCT5351522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolbutamide-d9
CAS 1219794-57-6
Renhedsgrad noter HPLC
Molekylær formel C12 2H9 H9 N2 O3 S
Mængde 2.5 mg
Synonym Benzenesulfonamide','N-[(butyl-1,1,2,2,3,3,4,4,4-d9-amino)carbonyl]-4-methyl- (ACI), N-[(Butyl-1,1,2,2,3,3,4,4,4-d9-amino)carbonyl]-4-methylbenzenesulfonamide (ACI), N-[(Butyl-d9-amino)carbonyl]-4-methylbenzenesulfonamide, 1-(Butyl-d9)-3-(4-methylphenyl)sulfonylurea, N-(Butyl-d9)-N'-(p-tolylsulfonyl)urea, Tolbutamide-d9, Tolbutamide-D9 (butyl-D9)
InChI formel InChI=1S/C12H18N2O3S/c1-3-4-9-13-12(15)14-18(16,17)11-7-5-10(2)6-8-11/h5-8H,3-4,9H2,1-2H3,(H2,13,14,15)/i1D3,3D2,4D2,9D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])NC(=O)NS(=O)(=O)c1ccc(C)cc1
IUPAC navn 1-(4-methylphenyl)sulfonyl-3-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)urea
Molekylvægt (g/mol) 279.4
Formel vægt 279.1603
Procent renhed >95