Få mere at vide
Tolazamide-d7, TRC
Artikelnummer. 30452375
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30452375 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452375 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53450225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tolazamide-d7
CAS 1794811-72-5
Molekylær formel C14H14D7N3O3S
Mængde 25 mg
Synonym N-[[(Hexahydro-1H-azepin-1-yl)amino]carbonyl]-4-methylbenzenesulfonamide-d7, 1-(4-Methylphenyl-d7-sulfonyl)-3-(hexahydro-1H-azepin-1-yl)urea, 1-(Hexahydro- 1-azepinyl)-3-p-tolylsulfonylurea-d7, Diabewas-d7, N-(p-Toluenesulfonyl)-N'-hexa- methyleniminourea-d7, NSC 70762, -d7 Norglycin-d7, Tolanase-d7, Tolinase-d7, U 17835-d7
InChI formel InChI=1S/C14H21N3O3S/c1-12-6-8-13(9-7-12)21(19,20)16-14(18)15-17-10-4-2-3-5-11-17/h6-9H,2-5,10-11H2,1H3,(H2,15,16,18)/i1D3,6D,7D,8D,9D
SMIL O=S(C1=C([2H])C([2H])=C(C([2H])([2H])[2H])C([2H])=C1[2H])(NC(NN2CCCCCC2)=O)=O
Molekylvægt (g/mol) 318.44
Formel vægt 318.1743