Få mere at vide
Tolan-d10, CDN
Artikelnummer. 30320448
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320448 0.1 g 0.1g
30318729 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320448 Leverandør CDN Leverandørnr. CDND20190.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diphenylacetylene-D10
CAS 19339-46-9
Molekylær formel C14 D10
Mængde 0.1 g
Synonym Benzene-1,2,3,4,5-d5, 6,6'-(1,2-ethynediyl)bis- (ACI), 6,6'-(1,2-Ethynediyl)bis[benzene-1,2,3,4,5-d5] (ACI), Acetylene, di(phenyl-d5)- (8 CI), Benzene-d5, 6,6'-(1,2-ethynediyl)bis- (9 CI), Diphenylacetylene-d10, Tolan-d10, Diphenylacetylene-D10
InChI formel InChI=1 S/C14H10/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-10 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D
SMIL [2 H]c1c([2 H])c([2 H])c(C#Cc2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-[2-(2,3,4,5,6-pentadeuteriophenyl)ethynyl]benzene
Molekylvægt (g/mol) 188.291
Formel vægt 188.141 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity