Learn More
Togni Reagent II, TRC
Specifications
Specifications
| Chemical Name or Material | Togni Reagent II |
| CAS | 887144-94-7 |
| Molecular Formula | C8 H4 F3 I O2 |
| Quantity | 5 g |
| Synonym | 1-(Trifluoromethyl)-1,2-benziodoxol-3(1H)-one |
| InChI Formula | InChI=1S/C8H4F3IO2/c9-8(10,11)12-6-4-2-1-3-5(6)7(13)14-12/h1-4H |
| SMILES | FC(F)(F)I1OC(=O)c2ccccc12 |
| IUPAC Name | 1-(trifluoromethyl)-1λ^{3},2-benziodoxol-3-one |
| Molecular Weight (g/mol) | 316.02 |
| Formula Weight | 315.9208 |