Få mere at vide
Tocris Bioscience™ Tocriscreen GPCR Compound Library
Mængde:
1 Set
Beskrivelse
Tocriscreen GPCR Compound Library is a library of 240 compounds (100 μl, 10 mM) for GPCR research. Contains compounds that target >30 types of GPCR, including peptide receptors, 5-HT receptors, chemokine receptors, adenosine receptors, adrenergic receptors and more. Adapted for popular techniques in drug discovery and cell biology research, including high throughput screening, high content screening, target validation and assay development.If this library does not suit your needs, please submit your requirements through our Tocriscreen PRO custom compound library service .
Tekniske data
Tekniske data
| Mængde | 1 Set |
| Mål | Screening Libraries |