Få mere at vide
Tocris Bioscience™ Psalmotoxin 1
Potent and selective ASIC1a channel blocker
Brand: Tocris Bioscience™ 5042/100U
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Psalmotoxin 1 is a potent and selective acid-sensing ion channel 1a (ASIC1a) blocker (IC50 = 0.9 nM). Displays no effect at ASIC1b, ASIC2a, ASIC3, heteromeric ASIC channels, ENaC and KV2.1/2.2/4.2/4.3 channels expressed in oocytes, at concentrations up to 100 nM. Displays potent analgesic properties against thermal, mechanical, chemical, inflammatory and neuropathic pain in rodents.Tekniske data
| 316808-68-1 | |
| 100 μg |