Få mere at vide
Tocris Bioscience™ GsMTx4
Beskrivelse
GsMTx4 is a spider venom peptide that selectively inhibits cationic mechanosensitive channels (MSCs) such as TRPC1 and TRPC6 and Piezo channels. GsMTx4 blocks stretch-activated cation channels in astrocytes, cardiac cells, and smooth and skeletal muscle cells. GsMTx4 also inhibits TACAN, a mechanosensitive ion channel involved in the pain response. GsMTx4 decreases the leptin-induced AMPK and MLC-2 phosphorylation in breast epithelial cells. GsMTx4 is neuroprotective and inhibits lysophosphatidylcholine-induced astrocyte toxicity in vivo in mice. GsMTx4 suppresses neurogenesis and enhances astrogenesis in human neural stem cells.
Tekniske data
Tekniske data
| CAS | 1209500-46-8 |
| Mængde | 100 μg |