Få mere at vide
Tocris Bioscience™ Decanoyl-RVKR-CMK
Artikelnummer. 18085062
Mængde:
1 mg
Artikelnummer. 18085062
Brand: Tocris Bioscience™ 3501/1
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Subtilisin/Kex2p-like proprotein convertase inhibitor
Decanoyl-RVKR-CMK is a subtilisin/Kex2p-like proprotein convertase inhibitor; blocks activity of all seven convertases (PC1, PC2, PC4, PACE4, PC5, PC7 and furin). Abolishes proET-1 processing in endothelial cells; inhibits regulated secretion of the neuronal polypeptide VGF in PC12 cells. Inhibits cleavage of glycoprotein B of human cytomegalovirus. Also inhibits cleavage of SARS-CoV-2 spike protein by furin and blocks viral cell entry (IC50 = 57 nM in plaque reduction assay).